Anti-IL3 Mouse Monoclonal Antibody [clone: 3B11]

Lieferant: OriGene
ORIGCF500182EA 749 EUR
ORIGCF500182
Anti-IL3 Mouse Monoclonal Antibody [clone: 3B11]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR