Anti-cyp7b1 Mouse Monoclonal Antibody [clone: 7H10]

Lieferant: OriGene
ORIGCF500244EA 749 EUR
ORIGCF500244
Anti-cyp7b1 Mouse Monoclonal Antibody [clone: 7H10]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: cyp7b1
Clonality: Monoclonal
Clone: 7H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR