Anti-TASP1 Mouse Monoclonal Antibody [clone: 1H6]

Lieferant: OriGene
ORIGCF502257EA 749 EUR
ORIGCF502257
Anti-TASP1 Mouse Monoclonal Antibody [clone: 1H6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TASP1
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR