Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 1E10]
Lieferant: OriGene
ORIGCF502262EA 749 EUR
ORIGCF502262
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 1E10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: DNAJB1
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human