Anti-PDE10A Mouse Monoclonal Antibody [clone: 1B2]

Lieferant: OriGene
ORIGCF500635EA 749 EUR
ORIGCF500635
Anti-PDE10A Mouse Monoclonal Antibody [clone: 1B2]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: PDE10A
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR