Anti-FCGR2A Mouse Monoclonal Antibody [clone: 9G5]

Lieferant: OriGene
ORIGCF500652EA 749 EUR
ORIGCF500652
Anti-FCGR2A Mouse Monoclonal Antibody [clone: 9G5]
Antikörper
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FCGR2A
Clonality: Monoclonal
Clone: 9G5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR