Anti-FHL1 Mouse Monoclonal Antibody [clone: 1D6]

Lieferant: OriGene
ORIGCF501295EA 749 EUR
ORIGCF501295
Anti-FHL1 Mouse Monoclonal Antibody [clone: 1D6]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FHL1
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR