Anti-USP5 Mouse Monoclonal Antibody [clone: 1F9]

Lieferant: OriGene
ORIGCF501297EA 770 EUR
ORIGCF501297
Anti-USP5 Mouse Monoclonal Antibody [clone: 1F9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: USP5
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR