Anti-C9orf41 Mouse Monoclonal Antibody [clone: 6A2]

Lieferant: OriGene
ORIGCF501307EA 749 EUR
ORIGCF501307
Anti-C9orf41 Mouse Monoclonal Antibody [clone: 6A2]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: C9orf41
Clonality: Monoclonal
Clone: 6A2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR