Anti-HIBCH Mouse Monoclonal Antibody [clone: 3H5]

Lieferant: OriGene
ORIGCF501317EA 770 EUR
ORIGCF501317
Anti-HIBCH Mouse Monoclonal Antibody [clone: 3H5]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR