Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 9A8]
Lieferant: OriGene
ORIGCF500308EA 749 EUR
ORIGCF500308
Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 9A8]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg
Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 9A8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human