Anti-RNF144B Mouse Monoclonal Antibody [clone: 11F4]

Lieferant: OriGene
ORIGCF500699EA 749 EUR
ORIGCF500699
Anti-RNF144B Mouse Monoclonal Antibody [clone: 11F4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: 11F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR