Anti-TBXAS1 Mouse Monoclonal Antibody [clone: 1A12]

Lieferant: OriGene
ORIGCF501365EA 749 EUR
ORIGCF501365
Anti-TBXAS1 Mouse Monoclonal Antibody [clone: 1A12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000 Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TBXAS1
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR