Anti-TRPM4 Mouse Monoclonal Antibody [clone: 3C4]

Lieferant: OriGene
ORIGCF500370EA 749 EUR
ORIGCF500370
Anti-TRPM4 Mouse Monoclonal Antibody [clone: 3C4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TRPM4
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR