Anti-CHCHD5 Mouse Monoclonal Antibody [clone: 4B3]

Lieferant: OriGene
ORIGCF502350EA 749 EUR
ORIGCF502350
Anti-CHCHD5 Mouse Monoclonal Antibody [clone: 4B3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CHCHD5
Clonality: Monoclonal
Clone: 4B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR