Anti-L1CAM Mouse Monoclonal Antibody [clone: 2C7]

Lieferant: OriGene
ORIGCF500729EA 749 EUR
ORIGCF500729
Anti-L1CAM Mouse Monoclonal Antibody [clone: 2C7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR