Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 4G2]

Lieferant: OriGene
ORIGCF500746EA 749 EUR
ORIGCF500746
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 4G2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50 Flow Cytometry: 1:100

Type: Primary
Antigen: AKR1A1
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR