Anti-C17orf28 Mouse Monoclonal Antibody [clone: 2E9]

Lieferant: OriGene
ORIGCF501372EA 749 EUR
ORIGCF501372
Anti-C17orf28 Mouse Monoclonal Antibody [clone: 2E9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C17orf28
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR