Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1H2]

Lieferant: OriGene
ORIGCF501388EA 749 EUR
ORIGCF501388
Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1H2]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100; Immunochemistry: 1:150

Type: Primary
Antigen: MOBKL1A
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR