Anti-KBTBD7 Mouse Monoclonal Antibody [clone: 1G3]

Lieferant: OriGene
ORIGCF501391EA 770 EUR
ORIGCF501391
Anti-KBTBD7 Mouse Monoclonal Antibody [clone: 1G3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: KBTBD7
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR