Anti-PLK1 Mouse Monoclonal Antibody [clone: 8C12]

Lieferant: OriGene
ORIGCF500393EA 749 EUR
ORIGCF500393
Anti-PLK1 Mouse Monoclonal Antibody [clone: 8C12]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone: 8C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR