Anti-SILV Mouse Monoclonal Antibody [clone: 7E3]

Lieferant: OriGene
ORIGCF500425EA 749 EUR
ORIGCF500425
Anti-SILV Mouse Monoclonal Antibody [clone: 7E3]
Antikörper
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: 7E3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR