Anti-SSB Mouse Monoclonal Antibody [clone: 2C8]

Lieferant: OriGene
ORIGCF500426EA 749 EUR
ORIGCF500426
Anti-SSB Mouse Monoclonal Antibody [clone: 2C8]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SSB
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR