Anti-BIRC7 Mouse Monoclonal Antibody [clone: 1B10]

Lieferant: OriGene
ORIGCF500761EA 749 EUR
ORIGCF500761
Anti-BIRC7 Mouse Monoclonal Antibody [clone: 1B10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: BIRC7
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR