Anti-SORD Mouse Monoclonal Antibody [clone: 9G9]

Lieferant: OriGene
ORIGCF500785EA 749 EUR
ORIGCF500785
Anti-SORD Mouse Monoclonal Antibody [clone: 9G9]
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SORD
Clonality: Monoclonal
Clone: 9G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR