Anti-PKLR Mouse Monoclonal Antibody [clone: 1A5]

Lieferant: OriGene
ORIGCF501411EA 749 EUR
ORIGCF501411
Anti-PKLR Mouse Monoclonal Antibody [clone: 1A5]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PKLR
Clonality: Monoclonal
Clone: 1A5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR