Anti-PKLR Mouse Monoclonal Antibody [clone: 1H4]

Lieferant: OriGene
ORIGCF501413EA 749 EUR
ORIGCF501413
Anti-PKLR Mouse Monoclonal Antibody [clone: 1H4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PKLR
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR