Anti-MIOX Mouse Monoclonal Antibody [clone: 2F4]

Lieferant: OriGene
ORIGCF501423EA 770 EUR
ORIGCF501423
Anti-MIOX Mouse Monoclonal Antibody [clone: 2F4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MIOX
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR