Anti-L1CAM Mouse Monoclonal Antibody [clone: 2G9]

Lieferant: OriGene
ORIGCF501441EA 749 EUR
ORIGCF501441
Anti-L1CAM Mouse Monoclonal Antibody [clone: 2G9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR