Anti-BRAF Mouse Monoclonal Antibody [clone: 1G8]

Lieferant: OriGene
ORIGCF500447EA 749 EUR
ORIGCF500447
Anti-BRAF Mouse Monoclonal Antibody [clone: 1G8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR