Anti-MAPK1 Mouse Monoclonal Antibody [clone: 6E5]

Lieferant: OriGene
ORIGCF500472EA 749 EUR
ORIGCF500472
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 6E5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 6E5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR