Anti-CD4 Mouse Monoclonal Antibody [clone: 6E10]

Lieferant: OriGene
ORIGCF500481EA 770 EUR
ORIGCF500481
Anti-CD4 Mouse Monoclonal Antibody [clone: 6E10]
Antikörper
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 6E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR