Anti-AKT2 Mouse Monoclonal Antibody [clone: 4H7]

Lieferant: OriGene
ORIGCF500814EA 749 EUR
ORIGCF500814
Anti-AKT2 Mouse Monoclonal Antibody [clone: 4H7]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 4H7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR