Anti-LEMD3 Mouse Monoclonal Antibody [clone: 3C8]

Lieferant: OriGene
ORIGCF500815EA 770 EUR
ORIGCF500815
Anti-LEMD3 Mouse Monoclonal Antibody [clone: 3C8]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: LEMD3
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR