Anti-SOD1 Mouse Monoclonal Antibody [clone: 8B10]

Lieferant: OriGene
ORIGCF500495EA 749 EUR
ORIGCF500495
Anti-SOD1 Mouse Monoclonal Antibody [clone: 8B10]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: SOD1
Clonality: Monoclonal
Clone: 8B10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR