Anti-MAPK12 Mouse Monoclonal Antibody [clone: 10E1]

Lieferant: OriGene
ORIGCF500500EA 749 EUR
ORIGCF500500
Anti-MAPK12 Mouse Monoclonal Antibody [clone: 10E1]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPK12
Clonality: Monoclonal
Clone: 10E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR