Anti-EIF2B3 Mouse Monoclonal Antibody [clone: 3H3]

Lieferant: OriGene
ORIGCF503681EA 749 EUR
ORIGCF503681
Anti-EIF2B3 Mouse Monoclonal Antibody [clone: 3H3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EIF2B3
Clonality: Monoclonal
Clone: 3H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR