Anti-PARVB Mouse Monoclonal Antibody [clone: 2H1]

Lieferant: OriGene
ORIGCF505678EA 770 EUR
ORIGCF505678
Anti-PARVB Mouse Monoclonal Antibody [clone: 2H1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PARVB
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR