Anti-NUBPL Mouse Monoclonal Antibody [clone: 3D11]

Lieferant: OriGene
ORIGCF503745EA 749 EUR
ORIGCF503745
Anti-NUBPL Mouse Monoclonal Antibody [clone: 3D11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NUBPL
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR