Anti-CST3 Mouse Monoclonal Antibody [clone: 7B8]

Lieferant: OriGene
ORIGCF504506EA 749 EUR
ORIGCF504506
Anti-CST3 Mouse Monoclonal Antibody [clone: 7B8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CST3
Clonality: Monoclonal
Clone: 7B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR