Anti-VBP1 Mouse Monoclonal Antibody [clone: 2G2]

Lieferant: OriGene
ORIGCF504911EA 749 EUR
ORIGCF504911
Anti-VBP1 Mouse Monoclonal Antibody [clone: 2G2]
Antikörper
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150

Type: Primary
Antigen: VBP1
Clonality: Monoclonal
Clone: 2G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR