Anti-ANXA7 Mouse Monoclonal Antibody [clone: 1C3]

Lieferant: OriGene
ORIGCF504925EA 749 EUR
ORIGCF504925
Anti-ANXA7 Mouse Monoclonal Antibody [clone: 1C3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ANXA7
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR