Anti-TRMU Mouse Monoclonal Antibody [clone: 2A4]

Lieferant: OriGene
ORIGCF505699EA 749 EUR
ORIGCF505699
Anti-TRMU Mouse Monoclonal Antibody [clone: 2A4]
Antikörper
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: TRMU
Clonality: Monoclonal
Clone: 2A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR