Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 4B3]

Lieferant: OriGene
ORIGCF505701EA 749 EUR
ORIGCF505701
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 4B3]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 4B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR