Anti-LPL Mouse Monoclonal Antibody [clone: 3C8]
Lieferant: OriGene
ORIGCF503785EA 749 EUR
ORIGCF503785
Anti-LPL Mouse Monoclonal Antibody [clone: 3C8]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150
Type: Primary
Antigen: LPL
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human