Anti-LPL Mouse Monoclonal Antibody [clone: 2G8]

Lieferant: OriGene
ORIGCF503790EA 749 EUR
ORIGCF503790
Anti-LPL Mouse Monoclonal Antibody [clone: 2G8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LPL
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR