Anti-HMOX2 Mouse Monoclonal Antibody [clone: 1B4]

Lieferant: OriGene
ORIGCF503822EA 770 EUR
ORIGCF503822
Anti-HMOX2 Mouse Monoclonal Antibody [clone: 1B4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: HMOX2
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR