Anti-MIF4GD Mouse Monoclonal Antibody [clone: 2H9]

Lieferant: OriGene
ORIGCF504558EA 749 EUR
ORIGCF504558
Anti-MIF4GD Mouse Monoclonal Antibody [clone: 2H9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MIF4GD
Clonality: Monoclonal
Clone: 2H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR