Anti-ATIC Mouse Monoclonal Antibody [clone: 4A10]

Lieferant: OriGene
ORIGCF504570EA 749 EUR
ORIGCF504570
Anti-ATIC Mouse Monoclonal Antibody [clone: 4A10]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: ATIC
Clonality: Monoclonal
Clone: 4A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR