Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1D7]

Lieferant: OriGene
ORIGCF504583EA 749 EUR
ORIGCF504583
Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1D7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: C17orf37
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR